Assay Details
Gene Symbol: Cd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022269.2 | NP_071605.2 | 4 - 5 | 637 | 147 | |
XM_006249717.2 | XP_006249779.1 | 4 - 5 | 659 | 147 | ||
XM_006249719.2 | XP_006249781.1 | 4 - 5 | 657 | 147 | ||
XM_006249723.2 | XP_006249785.1 | 4 - 5 | 665 | 147 | ||
XM_008769497.1 | XP_008767719.1 | 4 - 5 | 659 | 147 | ||
XM_008769498.1 | XP_008767720.1 | 4 - 5 | 659 | 147 | ||
GenBank mRNA | AB026900.1 | - | 4 - 5 | 644 | 147 | |
AB026901.1 | - | 4 - 5 | 581 | 147 | ||
AB026902.1 | - | 4 - 5 | 644 | 147 | ||
AB026903.1 | - | 4 - 5 | 581 | 147 | ||
AB026904.1 | - | 3 - 4 | 430 | 147 | ||
AF039583.1 | - | 4 - 5 | 578 | 147 | ||
AF039584.1 | - | 4 - 5 | 578 | 147 | ||
BC061869.1 | - | 4 - 5 | 601 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
