Assay Details
Gene Symbol: Thoc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024384.2 | NP_077360.2 | 3 - 4 | 460 | 82 | |
GenBank mRNA | BC079149.1 | - | 3 - 4 | 460 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
