Assay Details
Gene Symbol: Fez2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053600.2 | NP_446052.2 | 1 - 2 | 266 | 80 | |
XM_006239637.2 | XP_006239699.1 | 1 - 2 | 317 | 80 | ||
XM_006239638.2 | XP_006239700.1 | 1 - 2 | 317 | 80 | ||
XM_006239639.2 | XP_006239701.1 | 1 - 2 | 317 | 80 | ||
GenBank mRNA | AB076183.1 | - | 1 - 2 | 266 | 80 | |
AB080086.1 | - | 1 - 2 | 245 | 80 | ||
BC100060.1 | - | 1 - 2 | 247 | 80 | ||
CK359698.1 | - | 1 - 2 | 233 | 80 | ||
U64689.1 | - | 1 - 2 | 92 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
