Assay Details
Gene Symbol: Chst15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173310.3 | NP_775432.3 | 2 - 3 | 899 | 97 | |
XM_006230430.2 | XP_006230492.1 | 3 - 4 | 2466 | 97 | ||
XM_006230431.2 | XP_006230493.1 | 3 - 4 | 3206 | 97 | ||
XM_006230433.2 | XP_006230495.1 | 3 - 4 | 2701 | 97 | ||
XM_008759868.1 | XP_008758090.1 | 3 - 4 | 1462 | 97 | ||
XM_008759869.1 | XP_008758091.1 | 3 - 4 | 3205 | 97 | ||
GenBank mRNA | AB086953.1 | - | 2 - 3 | 899 | 97 | |
BC085687.1 | - | 2 - 3 | 1160 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
