Assay Details
Gene Symbol: Eef1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175838.1 | NP_787032.1 | 2 - 3 | 214 | 88 | |
GenBank mRNA | BC063162.1 | - | 2 - 3 | 210 | 88 | |
BC072542.1 | - | 2 - 3 | 209 | 88 | ||
BC091297.1 | - | 2 - 3 | 195 | 88 | ||
BC111707.1 | - | 1 - 2 | 178 | 88 | ||
BC128723.1 | - | 2 - 3 | 189 | 88 | ||
L10339.1 | - | 1 - 2 | 149 | 88 | ||
X61043.1 | - | 2 - 3 | 214 | 88 | ||
X63561.1 | - | 1 - 2 | 187 | 88 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
