Assay Details
Gene Symbol: Mrps28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047909.1 | NP_001041374.1 | 1 - 2 | 206 | 141 | |
GenBank mRNA | AY383686.1 | - | 1 - 2 | 206 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
