Assay Details
Gene Symbol: Mdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108099.1 | NP_001101569.1 | 11 | 1478 | 60 | |
XM_006241382.1 | XP_006241444.1 | 12 | 1843 | 60 | ||
XM_006241383.1 | XP_006241445.1 | 12 | 1407 | 60 | ||
XM_008765396.1 | XP_008763618.1 | 13 | 1915 | 60 | ||
XM_008765397.1 | XP_008763619.1 | 13 | 1479 | 60 | ||
XM_008765399.1 | XP_008763621.1 | 9 | 1225 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
ubiquitin-dependent protein catabolic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
traversing start control point of mitotic cell cycle
response to xenobiotic stimulus
response to toxic substance
response to carbohydrate
response to iron ion
positive regulation of gene expression
negative regulation of gene expression
negative regulation of protein processing
protein ubiquitination
peptidyl-lysine modification
protein catabolic process
protein destabilization
response to magnesium ion
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
protein localization to nucleus
regulation of protein catabolic process
response to cocaine
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to morphine
negative regulation of DNA damage response, signal transduction by p53 class mediator
establishment of protein localization
response to ether
positive regulation of cell cycle
negative regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
response to antibiotic
positive regulation of protein export from nucleus
response to steroid hormone
protein-containing complex assembly
cellular response to hydrogen peroxide
cellular response to antibiotic
cellular response to vitamin B1
cellular response to organic substance
cellular response to alkaloid
cellular response to growth factor stimulus
cellular response to peptide hormone stimulus
cellular response to estrogen stimulus
cellular response to organic cyclic compound
cellular response to hypoxia
cellular response to UV-C
negative regulation of signal transduction by p53 class mediator
p53 binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
enzyme binding
ubiquitin protein ligase binding
identical protein binding
peroxisome proliferator activated receptor binding
ubiquitin protein ligase activity
scaffold protein binding
negative regulation of transcription by RNA polymerase II
ubiquitin-dependent protein catabolic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
traversing start control point of mitotic cell cycle
response to xenobiotic stimulus
response to toxic substance
response to carbohydrate
response to iron ion
positive regulation of gene expression
negative regulation of gene expression
negative regulation of protein processing
protein ubiquitination
peptidyl-lysine modification
protein catabolic process
protein destabilization
response to magnesium ion
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
protein localization to nucleus
regulation of protein catabolic process
response to cocaine
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to morphine
negative regulation of DNA damage response, signal transduction by p53 class mediator
establishment of protein localization
response to ether
positive regulation of cell cycle
negative regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
response to antibiotic
positive regulation of protein export from nucleus
response to steroid hormone
protein-containing complex assembly
cellular response to hydrogen peroxide
cellular response to antibiotic
cellular response to vitamin B1
cellular response to organic substance
cellular response to alkaloid
cellular response to growth factor stimulus
cellular response to peptide hormone stimulus
cellular response to estrogen stimulus
cellular response to organic cyclic compound
cellular response to hypoxia
cellular response to UV-C
negative regulation of signal transduction by p53 class mediator
p53 binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
enzyme binding
ubiquitin protein ligase binding
identical protein binding
peroxisome proliferator activated receptor binding
ubiquitin protein ligase activity
scaffold protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P16885 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P634946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122640 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P634946 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P16885 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P16885 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P634946 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P16885 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P634946 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P634946 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16885 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91798 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10902573 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902575 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902576 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902566 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902570 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902567 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902569 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902572 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902571 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902565 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902568 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10902564 | - | - |
Rat Genome 230 2.0 Array | 1383288_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383485_PM_at | - | - |
Rat Genome 230 2.0 Array | 1384427_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383288_at | - | - |
Rat Genome 230 2.0 Array | 1383485_at | - | - |
Rat Genome 230 2.0 Array | 1384427_at | - | - |
Affymetrix HT Rat Focus Array | 1383288_at | - | - |
Affymetrix HT Rat Focus Array | 1384427_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1384427_at | - | - |
Affymetrix HT Rat Focus Array | 1383485_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383485_at | - | - |
Affymetrix HT Rat Focus Array | 1383288_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700021534.rn.1 | TC0700002764.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021533.rn.1 | TC0700002764.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021532.rn.1 | TC0700002764.rn.1 | RTA TC |
Rat Expression Set 230 | 1383485_PM_at | - | - |
Rat Expression Set 230 | 1384427_PM_at | - | - |
Rat Expression Set 230 | 1384427_at | - | - |
Rat Expression Set 230 | 1383485_at | - | - |
Rat Expression Set 230 | 1383288_PM_at | - | - |
Rat Expression Set 230 | 1383288_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902564 | - | - |
Rat Genome U34 Set | AA108308_i_at | - | - |
Rat Genome U34 Set | AA108308_s_at | - | - |
Rat Genome U34 Set | rc_AI639488_at | - | - |
Rat Genome U34 Set | AA875509_at | - | - |
Rat Genome U34 Set | rc_AA849796_at | - | - |
Rat Genome U34 Set | AA875509_at | - | - |
Rat Genome U34 Set | AA108308_i_at | - | - |
Rat Genome U34 Set | AA108308_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22250453 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364670 | - | - |
