Assay Details
Gene Symbol: Ndufb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106646.1 | NP_001100116.1 | 1 - 2 | 270 | 100 | |
GenBank mRNA | FQ216998.1 | - | 1 - 2 | 197 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
