Assay Details
Gene Symbol: Ndufb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106646.1 | NP_001100116.1 | 3 - 4 | 413 | 118 | |
GenBank mRNA | FQ216998.1 | - | 3 - 4 | 340 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
