Assay Details
Gene Symbol: Cd34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107202.2 | NP_001100672.1 | 7 - 8 | 971 | 71 | |
GenBank mRNA | EU448292.1 | - | 7 - 8 | 971 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
