Assay Details
Gene Symbol: Nudt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF188996.1 | - | 1 - 2 | 239 | 113 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | fibroblast growth factor 2 | Fgf2 |
Fgf-2, bFGF |
-4.7kb | Anti-sense | Chr. 2 - 124081072 - 124134133 on NCBI Build Rnor_6.0 |
Downstream | spermatogenesis associated 5 | Spata5 | 251.0 | Anti-sense | Chr. 2 - 124149872 - 124341663 on NCBI Build Rnor_6.0 |

More Information
Set Membership: |
![]() ![]() ![]() |
