Assay Details
Gene Symbol: Tbc1d25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106955.2 | NP_001100425.1 | 4 - 5 | 569 | 63 | |
XM_006256701.2 | XP_006256763.1 | 4 - 5 | 663 | 63 | ||
GenBank mRNA | BC161862.1 | - | 4 - 5 | 569 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
