Assay Details
Gene Symbol: Uchl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110165.1 | NP_001103635.1 | 6 - 7 | 559 | 134 | |
GenBank mRNA | AB043959.1 | - | 6 - 7 | 484 | 134 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
