Assay Details
Gene Symbol: Pcgf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129882.1 | NP_001123354.1 | 3 - 4 | 514 | 85 | |
XM_006231311.2 | XP_006231373.1 | 3 - 4 | 607 | 85 | ||
XM_008760377.1 | XP_008758599.1 | 2 - 3 | 232 | 85 | ||
GenBank mRNA | BC167067.1 | - | 3 - 4 | 514 | 85 | |
FQ220230.1 | - | 1 - 2 | 105 | 85 | ||
FQ233837.1 | - | 3 - 4 | 448 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
