Assay Details
Gene Symbol: Pcgf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129882.1 | NP_001123354.1 | 5 - 6 | 627 | 143 | |
XM_006231311.2 | XP_006231373.1 | 5 - 6 | 720 | 143 | ||
XM_008760377.1 | XP_008758599.1 | 4 - 5 | 345 | 143 | ||
GenBank mRNA | BC167067.1 | - | 5 - 6 | 627 | 143 | |
FQ214233.1 | - | 5 - 6 | 608 | 143 | ||
FQ220230.1 | - | 3 - 4 | 218 | 143 | ||
FQ233837.1 | - | 5 - 6 | 561 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
