Assay Details
Gene Symbol: Ston2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135874.1 | NP_001129346.1 | 3 - 4 | 683 | 96 | |
XM_006240406.2 | XP_006240468.1 | 4 - 5 | 1180 | 96 | ||
XM_006240407.2 | XP_006240469.1 | 4 - 5 | 918 | 96 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
