Assay Details
Gene Symbol: Ston2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135874.1 | NP_001129346.1 | 5 - 6 | 2730 | 74 | |
XM_006240406.2 | XP_006240468.1 | 6 - 7 | 3227 | 74 | ||
XM_006240407.2 | XP_006240469.1 | 6 - 7 | 2965 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P373103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97111 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P373103 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P373103 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P373103 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P373103 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P549916 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10891451 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891448 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891449 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891452 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891450 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891446 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10891445 | - | - |
Rat Genome 230 2.0 Array | 1380981_at | - | - |
Rat Genome 230 2.0 Array | 1380981_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380981_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380981_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600020854.rn.1 | TC0600002947.rn.1 | RTA TC |
Rat Expression Set 230 | 1380981_at | - | - |
Rat Expression Set 230 | 1380981_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891445 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372835 | - | - |
