Assay Details
Gene Symbol: Vdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031354.1 | NP_112644.1 | 8 - 9 | 964 | 82 | |
XM_006251661.1 | XP_006251723.1 | 9 - 10 | 1085 | 82 | ||
GenBank mRNA | AB039663.1 | - | 8 - 9 | 964 | 82 | |
AF268468.1 | - | 7 - 8 | 800 | 82 | ||
BC063164.1 | - | 8 - 9 | 933 | 82 | ||
FQ217146.1 | - | 5 - 6 | 82 | |||
FQ219952.1 | - | 5 - 6 | 82 | |||
FQ228796.1 | - | 5 - 6 | 689 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
