Assay Details
Gene Symbol: Apom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019373.2 | NP_062246.1 | 4 - 5 | 527 | 101 | |
GenBank mRNA | AF207821.1 | - | 4 - 5 | 504 | 101 | |
BC089807.1 | - | 4 - 5 | 476 | 101 | ||
FQ210093.1 | - | 4 - 5 | 526 | 101 | ||
FQ219449.1 | - | 4 - 5 | 527 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
