Assay Details
Gene Symbol: Dram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173427.1 | NP_001166898.1 | 1 - 2 | 248 | 96 | |
GenBank mRNA | BC168648.1 | - | 1 - 2 | 248 | 96 | |
FQ211820.1 | - | 1 - 2 | 376 | 96 | ||
FQ223303.1 | - | 1 - 2 | 309 | 96 | ||
FQ229264.1 | - | 1 - 2 | 309 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
