Assay Details
Gene Symbol: Cdh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168630.1 | NP_001162101.1 | 4 - 5 | 807 | 82 | |
XM_006232076.2 | XP_006232138.1 | 6 - 7 | 1031 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
