Assay Details
Gene Symbol: Cdh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168630.1 | NP_001162101.1 | 9 - 10 | 1629 | 89 | |
XM_006232076.2 | XP_006232138.1 | 11 - 12 | 1853 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
