Assay Details
Gene Symbol: Trmt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033902.2 | NP_001029074.2 | 1 - 2 | 165 | 100 | |
GenBank mRNA | BC103656.1 | - | 1 - 2 | 213 | 100 | |
BC166503.1 | - | 1 - 2 | 165 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
