Assay Details
Gene Symbol: Trmt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033902.2 | NP_001029074.2 | 3 - 4 | 283 | 121 | |
GenBank mRNA | BC103656.1 | - | 3 - 4 | 331 | 121 | |
BC166503.1 | - | 3 - 4 | 283 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
