Assay Details
Gene Symbol: Trmt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033902.2 | NP_001029074.2 | 5 - 6 | 421 | 93 | |
XM_008761469.1 | XP_008759691.1 | 3 - 4 | 296 | 93 | ||
GenBank mRNA | BC103656.1 | - | 5 - 6 | 469 | 93 | |
BC166503.1 | - | 5 - 6 | 421 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
