Assay Details
Gene Symbol: Pdhx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044242.1 | NP_001037707.1 | 7 - 8 | 967 | 103 | |
GenBank mRNA | AY310145.1 | - | 7 - 8 | 967 | 103 | |
BC091292.1 | - | 4 - 5 | 604 | 103 | ||
FQ232219.1 | - | 994 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
