Assay Details
Gene Symbol: Tesk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031578.1 | NP_113766.1 | 1 - 2 | 1327 | 81 | |
GenBank mRNA | BC081773.1 | - | 1 - 2 | 923 | 81 | |
D50864.1 | - | 1 - 2 | 1327 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
