Assay Details
Gene Symbol: Tesk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031578.1 | NP_113766.1 | 4 - 5 | 1647 | 77 | |
GenBank mRNA | BC081773.1 | - | 4 - 5 | 1243 | 77 | |
D50864.1 | - | 4 - 5 | 1647 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
