Assay Details
Gene Symbol: Btnl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212489.1 | NP_997654.1 | 1657 | 184 | ||
XM_008772738.1 | XP_008770960.1 | 8 | 2049 | 184 |
Gene Symbol: LOC100911938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772767.1 | XP_008770989.1 | 1657 | 184 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
