Assay Details
Gene Symbol: Mir10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031814.1 | 1 | 57 | 106 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | homeo box B5 | Hoxb5 | RGD1562292 | Sense | Chr. 10 - 84150319 - 84191398 on NCBI Build Rnor_6.0 | |
Downstream | homeo box B4 | Hoxb4 | RGD1560113 | 1.3kb | Sense | Chr. 10 - 84167331 - 84169913 on NCBI Build Rnor_6.0 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
