Assay Details
Gene Symbol: Ptpn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177593.1 | NP_001171064.1 | 12 - 13 | 1634 | 88 | |
NM_013088.2 | NP_037220.2 | 12 - 13 | 1622 | 88 | ||
GenBank mRNA | D83016.1 | - | 6 - 7 | 671 | 88 | |
U05963.1 | - | 12 - 13 | 1565 | 88 | ||
U09307.1 | - | 12 - 13 | 1547 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
