Assay Details
Gene Symbol: Ptpn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177593.1 | NP_001171064.1 | 15 - 16 | 1987 | 99 | |
NM_013088.2 | NP_037220.2 | 15 - 16 | 1975 | 99 | ||
GenBank mRNA | U09307.1 | - | 15 - 16 | 1900 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage checkpoint signaling
protein dephosphorylation
lipid metabolic process
triglyceride metabolic process
epidermal growth factor receptor signaling pathway
integrin-mediated signaling pathway
axonogenesis
brain development
heart development
hormone-mediated signaling pathway
positive regulation of signal transduction
dephosphorylation
platelet formation
microvillus organization
abortive mitotic cell cycle
regulation of cell adhesion mediated by integrin
negative regulation of cell adhesion mediated by integrin
multicellular organism growth
organ growth
peptidyl-tyrosine dephosphorylation
megakaryocyte development
atrioventricular canal development
ERBB signaling pathway
regulation of multicellular organism growth
hormone metabolic process
glucose homeostasis
positive regulation of mitotic cell cycle
negative regulation of insulin secretion
regulation of protein export from nucleus
positive regulation of hormone secretion
negative regulation of hormone secretion
platelet-derived growth factor receptor signaling pathway
neurotrophin TRK receptor signaling pathway
ephrin receptor signaling pathway
multicellular organismal reproductive process
genitalia development
inner ear development
homeostasis of number of cells within a tissue
negative regulation of cortisol secretion
negative regulation of growth hormone secretion
face morphogenesis
intestinal epithelial cell migration
positive regulation of ERK1 and ERK2 cascade
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
non-membrane spanning protein tyrosine phosphatase activity
insulin receptor binding
protein binding
protein domain specific binding
receptor tyrosine kinase binding
D1 dopamine receptor binding
phospholipase binding
insulin receptor substrate binding
peptide hormone receptor binding
DNA damage checkpoint signaling
protein dephosphorylation
lipid metabolic process
triglyceride metabolic process
epidermal growth factor receptor signaling pathway
integrin-mediated signaling pathway
axonogenesis
brain development
heart development
hormone-mediated signaling pathway
positive regulation of signal transduction
dephosphorylation
platelet formation
microvillus organization
abortive mitotic cell cycle
regulation of cell adhesion mediated by integrin
negative regulation of cell adhesion mediated by integrin
multicellular organism growth
organ growth
peptidyl-tyrosine dephosphorylation
megakaryocyte development
atrioventricular canal development
ERBB signaling pathway
regulation of multicellular organism growth
hormone metabolic process
glucose homeostasis
positive regulation of mitotic cell cycle
negative regulation of insulin secretion
regulation of protein export from nucleus
positive regulation of hormone secretion
negative regulation of hormone secretion
platelet-derived growth factor receptor signaling pathway
neurotrophin TRK receptor signaling pathway
ephrin receptor signaling pathway
multicellular organismal reproductive process
genitalia development
inner ear development
homeostasis of number of cells within a tissue
negative regulation of cortisol secretion
negative regulation of growth hormone secretion
face morphogenesis
intestinal epithelial cell migration
positive regulation of ERK1 and ERK2 cascade
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
non-membrane spanning protein tyrosine phosphatase activity
insulin receptor binding
protein binding
protein domain specific binding
receptor tyrosine kinase binding
D1 dopamine receptor binding
phospholipase binding
insulin receptor substrate binding
peptide hormone receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P004151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128013 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P004151 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P004151 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P004151 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P317773 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P999869 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701749110 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3675 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46030 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94344 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7314 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701749110 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10758733 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758730 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758728 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758729 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758738 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758741 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758735 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758739 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758731 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758734 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758737 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758742 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758736 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758732 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758743 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758740 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10758727 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10758744 | - | - |
Rat Genome 230 2.0 Array | 1388379_PM_at | - | - |
Rat Genome 230 2.0 Array | 1387668_at | - | - |
Rat Genome 230 2.0 Array | 1388379_at | - | - |
Rat Genome 230 2.0 Array | 1387668_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388379_at | - | - |
Affymetrix HT Rat Focus Array | 1387668_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388379_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387668_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1200006125.rn.1 | TC1200000673.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200006123.rn.1 | TC1200000673.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200006124.rn.1 | TC1200000673.rn.1 | RTA TC |
Rat Expression Set 230 | 1387668_PM_at | - | - |
Rat Expression Set 230 | 1388379_PM_at | - | - |
Rat Expression Set 230 | 1388379_at | - | - |
Rat Expression Set 230 | 1387668_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758727 | - | - |
Rat Genome U34 Set | U57499_at | - | - |
Rat Genome U34 Set | rc_AI171525_at | - | - |
Rat Genome U34 Set | U57499_g_at | - | - |
Rat Genome U34 Set | rc_AI138070_at | - | - |
Rat Genome U34 Set | U09307_at | - | - |
Rat Genome U34 Set | rc_AI012725_s_at | - | - |
Rat Genome U34 Set | rc_AI172465_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21382371 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363069 | - | - |
RatRef-12 Expression BeadChip | ILMN_64422 | - | - |
