Assay Details
Gene Symbol: Mettl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191814.1 | NP_001178743.1 | 4 - 5 | 903 | 86 | |
XM_006245658.2 | XP_006245720.1 | 5 - 6 | 1561 | 86 | ||
XM_006245661.2 | XP_006245723.1 | 4 - 5 | 1499 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
