Assay Details
Gene Symbol: Nnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073089.1 | 1 - 2 | 195 | 91 | ||
XM_006235442.2 | XP_006235504.1 | 1 - 2 | 95 | 91 | ||
GenBank mRNA | DY315493.1 | - | 1 - 2 | 139 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | bladder cancer associated protein | Blcap | -1.8kb | Anti-sense | Chr. 3 - 154040165 - 154042096 on NCBI Build Rnor_6.0 | |
Downstream |
catenin, beta like 1 |
Ctnnbl1 | 167.9kb | Sense | Chr. 3 - 154212245 - 154373170 on NCBI Build Rnor_6.0 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
