Assay Details
Gene Symbol: Nnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073089.1 | 2 - 3 | 281 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | bladder cancer associated protein | Blcap | -2.2kb | Anti-sense | Chr. 3 - 154040165 - 154042096 on NCBI Build Rnor_6.0 | |
Downstream |
catenin, beta like 1 |
Ctnnbl1 | 167.2kb | Sense | Chr. 3 - 154212245 - 154373170 on NCBI Build Rnor_6.0 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
