Assay Details
Gene Symbol: Bod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073160.1 | 2 - 3 | 583 | 80 | ||
GenBank mRNA | FQ213681.1 | - | 2 - 3 | 375 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cytoplasmic polyadenylation element binding protein 4 | Cpeb4 | -214.8kb | Anti-sense | Chr. 10 - 15984520 - 16047209 on NCBI Build Rnor_6.0 | |
Downstream | stanniocalcin 2 | Stc2 | 278.4kb | Sense | Chr. 10 - 16542909 - 16552675 on NCBI Build Rnor_6.0 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
