Assay Details
Gene Symbol: Cpsf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014245.2 | NP_001014267.1 | 2 - 3 | 322 | 101 | |
XM_006231062.2 | XP_006231124.1 | 3 - 4 | 465 | 101 | ||
XM_008760238.1 | XP_008758460.1 | 3 - 4 | 396 | 101 | ||
XM_008760239.1 | XP_008758461.1 | 3 - 4 | 396 | 101 | ||
GenBank mRNA | BC083864.1 | - | 2 - 3 | 325 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
