Assay Details
Gene Symbol: Spg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106433.2 | NP_001099903.2 | 8 - 9 | 1846 | 99 | |
XM_006232365.2 | XP_006232427.1 | 8 - 9 | 2174 | 99 | ||
XM_006232366.2 | XP_006232428.1 | 8 - 9 | 2165 | 99 | ||
XM_006232367.2 | XP_006232429.1 | 8 - 9 | 1963 | 99 | ||
GenBank mRNA | BC099217.1 | - | 6 - 7 | 879 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
