Assay Details
Gene Symbol: Fxyd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270688.1 | NP_001257617.1 | 6 - 7 | 423 | 76 | |
NM_001270689.1 | NP_001257618.1 | 6 - 7 | 451 | 76 | ||
NM_021909.2 | NP_068709.2 | 6 - 7 | 527 | 76 | ||
GenBank mRNA | BC060571.1 | - | 6 - 7 | 415 | 76 | |
DV726876.1 | - | 2 - 3 | 193 | 76 | ||
FM122563.1 | - | 6 - 7 | 401 | 76 | ||
FQ220058.1 | - | 6 - 7 | 451 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
