Assay Details
Gene Symbol: Scn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271046.2 | NP_001257975.1 | 5 - 6 | 670 | 100 | |
NM_017288.2 | NP_058984.1 | 5 - 6 | 884 | 100 | ||
GenBank mRNA | FQ216374.1 | - | 5 - 6 | 670 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
