Assay Details
Gene Symbol: Tnfrsf11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271235.1 | NP_001258164.1 | 8 - 9 | 801 | 73 | |
XM_008769487.1 | XP_008767709.1 | 8 - 9 | 779 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
