Assay Details
Gene Symbol: Scube3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271362.1 | NP_001258291.1 | 15 - 16 | 1913 | 99 | |
XM_008772802.1 | XP_008771024.1 | 15 - 16 | 3926 | 99 | ||
XM_008772803.1 | XP_008771025.1 | 16 - 17 | 1983 | 99 | ||
XM_008772804.1 | XP_008771026.1 | 14 - 15 | 3878 | 99 | ||
XM_008772805.1 | XP_008771027.1 | 15 - 16 | 3926 | 99 | ||
XM_008772806.1 | XP_008771028.1 | 14 - 15 | 3878 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
