Assay Details
Gene Symbol: Scube3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271362.1 | NP_001258291.1 | 5 - 6 | 599 | 65 | |
XM_008772802.1 | XP_008771024.1 | 4 - 5 | 2564 | 65 | ||
XM_008772803.1 | XP_008771025.1 | 5 - 6 | 621 | 65 | ||
XM_008772804.1 | XP_008771026.1 | 4 - 5 | 2564 | 65 | ||
XM_008772805.1 | XP_008771027.1 | 4 - 5 | 2564 | 65 | ||
XM_008772806.1 | XP_008771028.1 | 4 - 5 | 2564 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
