Assay Details
Gene Symbol: Gabrr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017292.2 | NP_058988.1 | 4 - 5 | 683 | 101 | |
XM_008763610.1 | XP_008761832.1 | 6 - 7 | 669 | 101 | ||
XM_008763611.1 | XP_008761833.1 | 3 - 4 | 1615 | 101 | ||
GenBank mRNA | D38494.1 | - | 4 - 5 | 683 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
