Assay Details
Gene Symbol: Gabrr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017292.2 | NP_058988.1 | 5 - 6 | 758 | 73 | |
XM_008763610.1 | XP_008761832.1 | 7 - 8 | 744 | 73 | ||
XM_008763611.1 | XP_008761833.1 | 4 - 5 | 1690 | 73 | ||
GenBank mRNA | D38494.1 | - | 5 - 6 | 758 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
