Assay Details
Gene Symbol: Ndufv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031064.2 | NP_112326.1 | 2 - 3 | 168 | 111 | |
GenBank mRNA | BC058495.1 | - | 2 - 3 | 172 | 111 | |
M22756.1 | - | 2 - 3 | 93 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
