Assay Details
Gene Symbol: Ndufv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031064.2 | NP_112326.1 | 4 - 5 | 348 | 79 | |
GenBank mRNA | BC058495.1 | - | 4 - 5 | 352 | 79 | |
M22756.1 | - | 4 - 5 | 273 | 79 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
