Assay Details
Gene Symbol: Ndufv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031064.2 | NP_112326.1 | 6 - 7 | 627 | 92 | |
GenBank mRNA | BC058495.1 | - | 6 - 7 | 631 | 92 | |
FQ215131.1 | - | 3 - 4 | 300 | 92 | ||
M22756.1 | - | 6 - 7 | 552 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
