Assay Details
Gene Symbol: Rpl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053971.3 | NP_446423.2 | 4 - 5 | 559 | 108 | |
XM_006249377.1 | XP_006249439.1 | 5 - 6 | 578 | 108 | ||
XM_006249378.2 | XP_006249440.1 | 5 - 6 | 612 | 108 | ||
GenBank mRNA | BC061784.1 | - | 4 - 5 | 555 | 108 | |
BC078761.1 | - | 4 - 5 | 562 | 108 | ||
BC097295.1 | - | 4 - 5 | 563 | 108 | ||
FQ209419.1 | - | 3 - 4 | 237 | 108 | ||
FQ213055.1 | - | 3 - 4 | 237 | 108 | ||
FQ213078.1 | - | 3 - 4 | 237 | 108 | ||
FQ216758.1 | - | 3 - 4 | 237 | 108 | ||
FQ217744.1 | - | 3 - 4 | 237 | 108 | ||
FQ219956.1 | - | 3 - 4 | 237 | 108 | ||
FQ219982.1 | - | 3 - 4 | 237 | 108 | ||
FQ220180.1 | - | 3 - 4 | 237 | 108 | ||
FQ220346.1 | - | 3 - 4 | 237 | 108 | ||
FQ220362.1 | - | 3 - 4 | 237 | 108 | ||
FQ220471.1 | - | 3 - 4 | 237 | 108 | ||
FQ220678.1 | - | 3 - 4 | 237 | 108 | ||
FQ220699.1 | - | 3 - 4 | 237 | 108 | ||
FQ220887.1 | - | 3 - 4 | 237 | 108 | ||
FQ220959.1 | - | 3 - 4 | 237 | 108 | ||
FQ221615.1 | - | 3 - 4 | 237 | 108 | ||
FQ222080.1 | - | 3 - 4 | 237 | 108 | ||
FQ222331.1 | - | 3 - 4 | 237 | 108 | ||
FQ222465.1 | - | 3 - 4 | 237 | 108 | ||
FQ222658.1 | - | 3 - 4 | 237 | 108 | ||
FQ223301.1 | - | 3 - 4 | 237 | 108 | ||
FQ226632.1 | - | 3 - 4 | 238 | 108 | ||
FQ227060.1 | - | 3 - 4 | 237 | 108 | ||
FQ227100.1 | - | 3 - 4 | 237 | 108 | ||
FQ227955.1 | - | 3 - 4 | 237 | 108 | ||
FQ227974.1 | - | 3 - 4 | 237 | 108 | ||
FQ228665.1 | - | 3 - 4 | 237 | 108 | ||
FQ229345.1 | - | 3 - 4 | 237 | 108 | ||
FQ229355.1 | - | 3 - 4 | 237 | 108 | ||
FQ229484.1 | - | 5 - 6 | 577 | 108 | ||
FQ229571.1 | - | 3 - 4 | 237 | 108 | ||
FQ229624.1 | - | 3 - 4 | 237 | 108 | ||
FQ229675.1 | - | 3 - 4 | 237 | 108 | ||
FQ229700.1 | - | 3 - 4 | 237 | 108 | ||
FQ229820.1 | - | 3 - 4 | 237 | 108 | ||
FQ229854.1 | - | 3 - 4 | 237 | 108 | ||
FQ229857.1 | - | 3 - 4 | 237 | 108 | ||
FQ230171.1 | - | 3 - 4 | 237 | 108 | ||
FQ230222.1 | - | 3 - 4 | 237 | 108 | ||
FQ232778.1 | - | 3 - 4 | 237 | 108 | ||
FQ233198.1 | - | 3 - 4 | 237 | 108 | ||
FQ233475.1 | - | 3 - 4 | 237 | 108 | ||
FQ234880.1 | - | 5 - 6 | 576 | 108 | ||
FQ235095.1 | - | 3 - 4 | 237 | 108 | ||
X87107.1 | - | 5 - 6 | 573 | 108 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
