Assay Details
Gene Symbol: Oaz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297557.1 | NP_001284486.1 | 1 - 2 | 225 | 88 | |
NM_139081.2 | NP_620781.1 | 1 - 2 | 225 | 88 | ||
GenBank mRNA | D10706.1 | - | 1 - 2 | 197 | 88 | |
FQ211375.1 | - | 1 - 2 | 225 | 88 | ||
FQ213504.1 | - | 1 - 2 | 225 | 88 | ||
FQ213794.1 | - | 1 - 2 | 225 | 88 | ||
FQ214019.1 | - | 1 - 2 | 220 | 88 | ||
FQ214051.1 | - | 1 - 2 | 220 | 88 | ||
FQ214373.1 | - | 1 - 2 | 220 | 88 | ||
FQ214458.1 | - | 1 - 2 | 220 | 88 | ||
FQ216817.1 | - | 1 - 2 | 225 | 88 | ||
FQ218486.1 | - | 1 - 2 | 220 | 88 | ||
FQ218990.1 | - | 1 - 2 | 220 | 88 | ||
FQ220491.1 | - | 1 - 2 | 220 | 88 | ||
FQ220580.1 | - | 1 - 2 | 225 | 88 | ||
FQ225116.1 | - | 1 - 2 | 220 | 88 | ||
FQ225909.1 | - | 1 - 2 | 220 | 88 | ||
FQ225949.1 | - | 1 - 2 | 223 | 88 | ||
FQ229954.1 | - | 1 - 2 | 220 | 88 | ||
FQ230452.1 | - | 1 - 2 | 220 | 88 | ||
FQ231088.1 | - | 1 - 2 | 220 | 88 | ||
FQ232667.1 | - | 1 - 2 | 220 | 88 | ||
FQ232997.1 | - | 1 - 2 | 220 | 88 | ||
FQ233723.1 | - | 1 - 2 | 220 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001297557.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
